(3-Glycidoxypropyl)triethoxysilane ≥96.0%
Leverantör: TCI
|
Varning
|
Synonyms:
Geniosil GF 82, Glycidyl 3-Triethoxysilylpropyl ether, Triethoxy(3-glycidyloxypropyl)silane, (3-Glycidyloxypropyl)triethoxysilane, Triethoxy(3-(oxiran-2-ylmethoxy)propyl)silane, (3-(2,3-Epoxypropoxy)propyl)triethoxysilane
T2675-25G
T2675-5G
TCIAT2675-25GEA
415
SEK
TCIAT2675-25G
TCIAT2675-5G
(3-Glycidoxypropyl)triethoxysilane ≥96.0%
(3-Glycidoxypropyl)triethoxysilane
Formel:
C₁₂H₂₆O₅Si Kokpunkt: 135°C Flampunkt: 118°C |
MDL Number:
MFCD01311705 CAS nummer: 2602-34-8 EINECS: 220-011-6 |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...