5,6,7,8-Tetrahydronaphthalene-1-carboxylic acid ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

T2672-25G T2672-5G
TCIAT2672-25GEA 5175 SEK
TCIAT2672-25G TCIAT2672-5G
5,6,7,8-Tetrahydronaphthalene-1-carboxylic acid ≥98.0% (by GC, titration analysis)
5,6,7,8-Tetrahydronaphthalene-1-carboxylic acid
Formel: C₁₁H₁₂O₂
MW: 176,2118 g/mol
MDL Number: MFCD01464030
CAS nummer: 4242-18-6

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 150.0 to 154.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR