2,3,4,5-Tetramethoxytoluene ≥97.0%

Leverantör: TCI EUROPE

Synonyms: 1,2,3,4-Tetramethoxy-5-methylbenzene

T2626-25G T2626-5G
TCIAT2626-25GEA 1400 SEK
TCIAT2626-25G TCIAT2626-5G
2,3,4,5-Tetramethoxytoluene ≥97.0%
2,3,4,5-Tetramethoxytoluene
Formel: C₁₁H₁₆O₄
MW: 212,25 g/mol
Kokpunkt: 120°C
MDL Number: MFCD07370118
CAS nummer: 35896-58-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cdde814d9ca7c3d18f633b0e086deec662cabd9e&vid=cdde814d9ca7c3d18f633b0e086deec662cabd9e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR