2-Thiophenemethanethiol ≥95.0%

Leverantör: TCI

Synonyms: 2-(Mercaptomethyl)thiophene, Thenyl Mercaptan

T2613-1G T2613-5G
TCIAT2613-1GEA 630 SEK
TCIAT2613-1G TCIAT2613-5G
2-Thiophenemethanethiol ≥95.0%
2-Thiophenemethanethiol
Formel: C₅H₆S₂
Kokpunkt: 86°C
MDL Number: MFCD00107135
CAS nummer: 6258-63-5
EINECS: 228-394-1

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR