N-[2-(Trimethylsilyl)ethoxycarbonyloxy]succinimide ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
2,5-Dioxopyrrolidin-1-yl (2-(trimethylsilyl)ethyl) carbonate, Teoc-OSu
T2591-1G
T2591-5G
TCIAT2591-1GEA
485
SEK
TCIAT2591-1G
TCIAT2591-5G
N-[2-(Trimethylsilyl)ethoxycarbonyloxy]succinimide ≥98.0%
N-[2-(Trimethylsilyl)ethoxycarbonyloxy]succinimide
Formel:
C₁₀H₁₇NO₅Si MW: 259,33 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD02683467 CAS nummer: 78269-85-9 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 98.0 to 102.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...