(+)-Terpinen-4-ol ≥93.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
(+)-1-Isopropyl-4-methyl-3-cyclohexen-1-ol, (+)-4-Hydroxy-4-isopropyl-1-methyl-1-cyclohexene, (+)-p-Menth-1-en-4-ol
T2581-25G
T2581-5G
TCIAT2581-25GEA
1600
SEK
TCIAT2581-25G
TCIAT2581-5G
(+)-Terpinen-4-ol ≥93.0%
(+)-Terpinen-4-ol
Formel:
C₁₀H₁₈O MW: 154,25 g/mol Kokpunkt: 90°C Flampunkt: 79°C |
MDL Number:
MFCD00077431 CAS nummer: 2438-10-0 |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 93.0 % |
Enantiomeric ratio (GC) | (+) >65, (-) <35 |
Specific gravity (20/20) | 0.9320 to 0.9440 |
Refractive index n20/D | 1.4770 to 1.4810 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...