2,3,4,6-Tetra-O-acetyl-D-mannopyranosyl fluoride ≥95.0%
Leverantör: TCI EUROPE
T2567-1G
T2567-5G
TCIAT2567-1GEA
2850
SEK
TCIAT2567-1G
TCIAT2567-5G
2,3,4,6-Tetra-O-acetyl-D-mannopyranosyl fluoride ≥95.0%
2,3,4,6-Tetra-O-acetyl-D-mannopyranosyl fluoride
Formel:
C₁₄H₁₉FO₉ MW: 350,30 g/mol Storage Temperature: Kylskåp |
CAS nummer:
174511-17-2 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...