Tributyl(2-methoxyethyl)phosphonium bis(trifluoromethanesulphonyl)imide ≥98.0%

Leverantör: TCI EUROPE
Varning

T2564-5G T2564-25G
TCIAT2564-5GEA 1525 SEK
TCIAT2564-5G TCIAT2564-25G
Tributyl(2-methoxyethyl)phosphonium bis(trifluoromethanesulphonyl)imide ≥98.0%
Tributyl(2-methoxyethyl)phosphonium bis(trifluoromethanesulphonyl)imide
Formel: C₁₇H₃₄F₆NO₅PS₂
MW: 541,55 g/mol
MDL Number: MFCD11112190
CAS nummer: 959698-44-3

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (Ion exchange titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR