Tri-p-tolylamine ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 4,4',4''-Trimethyltriphenylamine

T2558-25G T2558-5G
TCIAT2558-25GEA 3875 SEK
TCIAT2558-25G TCIAT2558-5G
Tri-p-tolylamine ≥98.0%
Tri-p-tolylamine
Formel: C₂₁H₂₁N
MW: 287,41 g/mol
Kokpunkt: 170°C
MDL Number: MFCD00674043
CAS nummer: 1159-53-1
EINECS: 214-595-1

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 115.0 to 119.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR