(R)-(-)-Tetrahydrofurfurylamine ≥98.0% (by GC, titration analysis)
Leverantör: TCI
|
Varning
|
Synonyms:
(R)-(-)-2-(Aminomethyl)tetrahydrofuran, (R)-2-Tetrahydrofurfurylamine
T2551-1G
T2551-200MG
T2551-5G
TCIAT2551-1GEA
1750
SEK
TCIAT2551-1G
TCIAT2551-200MG
TCIAT2551-5G
(R)-(-)-Tetrahydrofurfurylamine ≥98.0% (by GC, titration analysis)
(R)-(-)-Tetrahydrofurfurylamine
Formel:
C₅H₁₁NO MW: 101.15 g/mol Kokpunkt: 60°C Flampunkt: 46°C |
MDL Number:
MFCD00192476 CAS nummer: 7202-43-9 UN: 2943 ADR: 3 III |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | -12.0 to -18.0 deg (C=2, MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...