2,2,2-Trifluoro-N-phenylacetimidoyl chloride ≥98.0% (by GC, titration analysis)

Leverantör: TCI
Fara

T2547-25G T2547-5G
TCIAT2547-25GEA 14150 SEK
TCIAT2547-25G TCIAT2547-5G
2,2,2-Trifluoro-N-phenylacetimidoyl chloride ≥98.0% (by GC, titration analysis)
2,2,2-Trifluoro-N-phenylacetimidoyl chloride
Formel: C₈H₅ClF₃N
Kokpunkt: 53°C
Storage Temperature: Kylskåp
MDL Number: MFCD11042687
CAS nummer: 61881-19-4
UN: 2924
ADR: 3,III

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR