2,3,5,6-Tetrafluoro-4-mercaptobenzoic acid ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

T2542-1G T2542-5G
TCIAT2542-1GEA 2150 SEK
TCIAT2542-1G TCIAT2542-5G
2,3,5,6-Tetrafluoro-4-mercaptobenzoic acid ≥98.0% (by GC, titration analysis)
2,3,5,6-Tetrafluoro-4-mercaptobenzoic acid
Formel: C₇H₂F₄O₂S
MW: 226,14 g/mol
CAS nummer: 5211-44-9

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 151.0 to 155.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR