Tetrafluorosuccinic anhydride ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

T2478-1G T2478-5G
TCIAT2478-1GEA 765 SEK
TCIAT2478-1G TCIAT2478-5G
Tetrafluorosuccinic anhydride ≥98.0% (by GC, titration analysis)
Tetrafluorosuccinic anhydride
Formel: C₄F₄O₃
MW: 172,04 g/mol
Kokpunkt: 55°C
Densitet: 1,62
Storage Temperature: Frys
MDL Number: MFCD00037917
CAS nummer: 699-30-9
EINECS: 211-832-0
UN: 1993
ADR: 3,II

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Methanolysis Method) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR