2,2,3,3-Tetramethylcyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

T2350-25G T2350-5G
TCIAT2350-25GEA 1525 SEK
TCIAT2350-25G TCIAT2350-5G
2,2,3,3-Tetramethylcyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)
2,2,3,3-Tetramethylcyclopropanecarboxylic acid
Formel: C₈H₁₄O₂
MW: 142,20 g/mol
MDL Number: MFCD00011538
CAS nummer: 15641-58-4

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 117.0 to 122.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR