Tris(4-nitrophenyl)amine ≥96.0%

Leverantör: TCI EUROPE
Varning

T2333-25G T2333-5G
TCIAT2333-25GEA 3525 SEK
TCIAT2333-25G TCIAT2333-5G
Tris(4-nitrophenyl)amine ≥96.0%
Tris(4-nitrophenyl)amine
Formel: C₁₈H₁₂N₄O₆
Smältpunkt: 280°C
MDL Number: MFCD00024636
CAS nummer: 20440-93-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f8065f0673f0c868ac2ecd832078694ac926bc9&vid=6f8065f0673f0c868ac2ecd832078694ac926bc9&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 96.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR