4,4,5,5-Tetramethyl-2-vinyl-1,3,2-dioxaborolane ≥93.0% stabilised

Leverantör: TCI
Varning


Stabilization: Stabilised with phenothiazine

T2297-1G T2297-25G T2297-5G
TCIAT2297-1GEA 325 SEK
TCIAT2297-1G TCIAT2297-25G TCIAT2297-5G
4,4,5,5-Tetramethyl-2-vinyl-1,3,2-dioxaborolane ≥93.0% stabilised
4,4,5,5-Tetramethyl-2-vinyl-1,3,2-dioxaborolane
Formel: C₈H₁₅BO₂
Kokpunkt: 40°C
Flampunkt: 34°C
Storage Temperature: Kylskåp
MDL Number: MFCD00192492
CAS nummer: 75927-49-0
UN: 3272
ADR: 3,III

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 93.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR