2,2,3,3-Tetrafluoropropyl acrylate ≥98.0% stabilised
Leverantör: TCI
|
Varning
|
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
T2240-25G
T2240-5G
TCIAT2240-25GEA
1600
SEK
TCIAT2240-25G
TCIAT2240-5G
2,2,3,3-Tetrafluoropropyl acrylate ≥98.0% stabilised
2,2,3,3-Tetrafluoropropyl acrylate
Formel:
C₆H₆F₄O₂ Kokpunkt: 75°C Storage Temperature: Kylskåp |
CAS nummer:
7383-71-3 EINECS: 230-957-1 UN: 3272 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...