9-(p-Tolyl)carbazole ≥98.0%
Leverantör: TCI
Varning
Synonyms: 9-(p-Tolyl)-9H-carbazole
T2227-1G T2227-5G
TCIAT2227-1GEA 605 SEK
TCIAT2227-1G TCIAT2227-5G
9-(p-Tolyl)carbazole ≥98.0%
9-(p-Tolyl)carbazole
Formel: C₁₉H₁₅N
MW: 257.34 g/mol
MDL Number: MFCD14708175
CAS nummer: 19264-73-4
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 105.0 to 109.0 deg-C