o-Tolyl-3,5-xylyl ether ≥97.0%
Leverantör: TCI EUROPE
Synonyms:
2,3',5'-Trimethyldiphenyl Ether
T2213-25G
T2213-5G
TCIAT2213-25GEA
5050
SEK
TCIAT2213-25G
TCIAT2213-5G
o-Tolyl-3,5-xylyl ether ≥97.0%
o-Tolyl-3,5-xylyl ether
Formel:
C₁₅H₁₆O MW: 212,29 g/mol |
CAS nummer:
196604-20-3 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...