Trisodium-N-(1-carboxylatoethyl)iminodiacetate hydrate ≥95.0%
Leverantör: TCI
Synonyms:
MGDA, Trisodium 2-Methylnitrilotriacetate, N-(1-Carboxylatoethyl)iminodiacetic Acid Trisodium Salt, N,N-Bis(carboxylatomethyl)alanine Trisodium Salt, Methyl Glycine Diacetic Acid Trisodium Salt
T2202-25G
T2202-500G
TCIAT2202-25GEA
230
SEK
TCIAT2202-25G
TCIAT2202-500G
Trisodium-N-(1-carboxylatoethyl)iminodiacetate hydrate ≥95.0%
Trisodium-N-(1-carboxylatoethyl)iminodiacetate hydrate
Formel:
C₇H₁₁NO₆ |
CAS nummer:
164462-16-2 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (Chelometric Titration) | min. 95.0 % (calcd.on anh.substance) |
Water | 5.0 to 13.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...