2,4,5-Trifluoro-1,3-phenylenediamine ≥98.0%

Leverantör: TCI
Varning

Synonyms: 1,3-Diamino-2,4,5-trifluorobenzene

T2060-1G T2060-5G
TCIAT2060-1GEA 1525 SEK
TCIAT2060-1G TCIAT2060-5G
2,4,5-Trifluoro-1,3-phenylenediamine ≥98.0%
2,4,5-Trifluoro-1,3-phenylenediamine
Formel: C₆H₅F₃N₂
MDL Number: MFCD04114298
CAS nummer: 321182-37-0

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 74.0 to 77.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR