2-(2-Thienyl)-1,3-dioxolane ≥97.0%

Leverantör: TCI EUROPE

Synonyms: 2-(1,3-Dioxolan-2-yl)thiophene

T2049-25G T2049-5G
TCIAT2049-25GEA 3800 SEK
TCIAT2049-25G TCIAT2049-5G
2-(2-Thienyl)-1,3-dioxolane ≥97.0%
2-(2-Thienyl)-1,3-dioxolane
Formel: C₇H₈O₂S
MW: 156,20 g/mol
Kokpunkt: 111°C
Storage Temperature: Kylskåp
MDL Number: MFCD01924576
CAS nummer: 58268-08-9

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR