Bis(carboxymethyl)trithiocarbonate ≥98.0%

Leverantör: TCI

Synonyms: Trithiocarbonic Acid Bis(carboxymethyl) Ester

T2036-25G T2036-5G
TCIAT2036-25GEA 7050 SEK
TCIAT2036-25G TCIAT2036-5G
Bis(carboxymethyl)trithiocarbonate ≥98.0%
Bis(carboxymethyl)trithiocarbonate
Formel: C₅H₆O₄S₃
MDL Number: MFCD00004357
CAS nummer: 6326-83-6
EINECS: 228-693-7

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 170.0 to 174.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR