Sodium trifluoromethanesulphinate ≥95.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Trifluoromethanesulphinic acid Sodium Salt, Langlois Reagent

T2033-25G T2033-5G
TCIAT2033-25GEA 2325 SEK
TCIAT2033-25G TCIAT2033-5G
Sodium trifluoromethanesulphinate ≥95.0%
Sodium trifluoromethanesulphinate
Formel: CHF₃O₂S
MW: 156,05 g/mol
MDL Number: MFCD03092989
CAS nummer: 2926-29-6

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (Sodium hypochlorite Method) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR