Dimethyl-5-bromoisophthalate ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Dimethyl 5-bromoisophthalate, 5-Bromoisophthalic acid dimethyl ester
H50629.03 H50629.06
H50629.03EA 875 SEK
H50629.03 H50629.06
Dimethyl-5-bromoisophthalate ≥98%
Dimethyl-5-bromoisophthalate
Purity: 98% CAS No.: 51760-21-5 Molecular Formula: C10H9BrO4 Formula Weight: 273.08 Melting Point: 88-90° MDL No.: MFCD00078709
Formel: C₁₀H₉BrO₄
MW: 273.08 g/mol
Smältpunkt: 88…90 °C
Storage Temperature: Omgivande
MDL Number: MFCD00078709
CAS nummer: 51760-21-5
EINECS: 257-386-0