Tris(4-iodophenyl)amine ≥97.0%

Leverantör: TCI
Varning

T1724-25G T1724-5G
TCIAT1724-25GEA 2800 SEK
TCIAT1724-25G TCIAT1724-5G
Tris(4-iodophenyl)amine ≥97.0%
Tris(4-iodophenyl)amine
Formel: C₁₈H₁₂I₃N
MDL Number: MFCD01321198
CAS nummer: 4181-20-8

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 187.0 to 191.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR