2-Hydroxyethyl methyl terephthalate ≥97.0%
Leverantör: TCI EUROPE
Synonyms:
Terephthalic Acid 2-Hydroxyethyl Methyl Ester
T1679-5G
T1679-1G
TCIAT1679-5GEA
3625
SEK
TCIAT1679-5G
TCIAT1679-1G
2-Hydroxyethyl methyl terephthalate ≥97.0%
2-Hydroxyethyl methyl terephthalate
Formel:
C₁₁H₁₂O₅ MW: 224,21 g/mol |
MDL Number:
MFCD01321191 CAS nummer: 3645-00-9 EINECS: 222-877-0 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 81.0 to 84.0 deg-C |
Solubility in hot Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...