N,N,O-Tris(trimethylsilyl)hydroxylamine ≥97.0%
Leverantör: TCI EUROPE
|
Fara
|
T1675-1G
T1675-5G
TCIAT1675-1GEA
1275
SEK
TCIAT1675-1G
TCIAT1675-5G
N,N,O-Tris(trimethylsilyl)hydroxylamine ≥97.0%
N,N,O-Tris(trimethylsilyl)hydroxylamine
Formel:
C₉H₂₇NOSi₃ MW: 249,58 g/mol Kokpunkt: 128°C Smältpunkt: –36°C Storage Temperature: Kylskåp |
MDL Number:
MFCD01321190 CAS nummer: 21023-20-1 UN: 1760 ADR: 8,III |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...