O-(Trimethylsilyl)hydroxylamine ≥95.0%

Leverantör: TCI EUROPE
Fara

T1669-25G T1669-5G
TCIAT1669-25GEA 7000 SEK
TCIAT1669-25G TCIAT1669-5G
O-(Trimethylsilyl)hydroxylamine ≥95.0%
O-(Trimethylsilyl)hydroxylamine
Formel: C₃H₁₁NOSi
MW: 105,21 g/mol
Kokpunkt: 105°C
Flampunkt: 12°C
Storage Temperature: Kylskåp
MDL Number: MFCD00010872
CAS nummer: 22737-36-6
UN: 2733
ADR: 3,II

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR