Triphenylsulphonium tetrafluoroborate ≥98.0%

Leverantör: TCI EUROPE
Fara

T1608-1G T1608-5G
TCIAT1608-1GEA 2050 SEK
TCIAT1608-1G TCIAT1608-5G
Triphenylsulphonium tetrafluoroborate ≥98.0%
Triphenylsulphonium tetrafluoroborate
Formel: C₁₈H₁₅BF₄S
MW: 350,18 g/mol
MDL Number: MFCD00671581
CAS nummer: 437-13-8
UN: 1759
ADR: 8,III

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 192.0 to 196.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR