Tetraethylene glycol diacrylate ≥90.0% stabilised
Leverantör: TCI EUROPE
Fara
Stabilization: Stabilised with 4-methoxyphenol (MEHQ)
T1569-25G T1569-500G
TCIAT1569-25GEA 215 SEK
TCIAT1569-25G TCIAT1569-500G
Tetraethylene glycol diacrylate ≥90.0% stabilised
Tetraethylene glycol diacrylate
Formel: C₁₄H₂₂O₇
MW: 302,32 g/mol
Flampunkt: 185°C
MDL Number: MFCD00008630
CAS nummer: 17831-71-9
EINECS: 241-789-3
UN: 1760
ADR: 8,III
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 90.0 %