Tetraisopropylthiuram disulphide ≥98.0%
Leverantör: TCI
|
Varning
|
Synonyms:
Bis(diisopropylthiocarbamoyl) Disulphide
T1555-25G
T1555-5G
TCIAT1555-25GEA
5875
SEK
TCIAT1555-25G
TCIAT1555-5G
Tetraisopropylthiuram disulphide ≥98.0%
Tetraisopropylthiuram disulphide
Formel:
C₁₄H₂₈N₂S₄ |
MDL Number:
MFCD00009860 CAS nummer: 4136-91-8 |
Specifikation testresultat
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 111.0 to 116.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...