3,3,5,5-Tetramethyl-1-pyrroline-N-oxide ≥98.0%
Leverantör: TCI EUROPE
Varning
T1554-1G T1554-100MG
TCIAT1554-1GEA 5025 SEK
TCIAT1554-1G TCIAT1554-100MG
3,3,5,5-Tetramethyl-1-pyrroline-N-oxide ≥98.0%
3,3,5,5-Tetramethyl-1-pyrroline-N-oxide
Formel: C₈H₁₅NO
MW: 141,21 g/mol
Kokpunkt: 73 °C
Smältpunkt: 61 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00010542
CAS nummer: 10135-38-3
Order Now
Specifikation testresultat
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 58.0 to 62.0 deg-C