2-(Trimethylsilyloxy)furan ≥96.0%
Leverantör: TCI
Fara
T1536-25G T1536-5G
TCIAT1536-25GEA 9500 SEK
TCIAT1536-25G TCIAT1536-5G
2-(Trimethylsilyloxy)furan ≥96.0%
2-(Trimethylsilyloxy)furan
Formel: C₇H₁₂O₂Si
Kokpunkt: 50°C
Flampunkt: 18°C
Storage Temperature: Kylskåp
MDL Number: MFCD00011729
CAS nummer: 61550-02-5
UN: 1993
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 96.0 %