2-Methoxyethyl-p-toluenesulphonate ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 2-Methoxyethyl 4-methylbenzenesulphonate, p-Toluenesulphonic acid 2-Methoxyethyl Ester

T1303-25G T1303-500G T1303-100G
TCIAT1303-25GEA 710 SEK
TCIAT1303-25G TCIAT1303-500G TCIAT1303-100G
2-Methoxyethyl-p-toluenesulphonate ≥98.0%
2-Methoxyethyl-p-toluenesulphonate
Formel: C₁₀H₁₄O₄S
MW: 230,28 g/mol
Kokpunkt: 141°C
Smältpunkt: 10°C
MDL Number: MFCD00025910
CAS nummer: 17178-10-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec821fcf0d8fa9c842a7a89265a53d80af10bfc6&vid=ec821fcf0d8fa9c842a7a89265a53d80af10bfc6&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR