N-(Trimethylsilyl)morpholine ≥97.0%
Leverantör: TCI
Fara
Synonyms: 4-(Trimethylsilyl)morpholine
T1277-25ML T1277-5ML
TCIAT1277-25MLEA 1625 SEK
TCIAT1277-25ML TCIAT1277-5ML
N-(Trimethylsilyl)morpholine ≥97.0%
N-(Trimethylsilyl)morpholine
Formel: C₇H₁₇NOSi
Kokpunkt: 102°C
Flampunkt: 29°C
Storage Temperature: Kylskåp
MDL Number: MFCD00010098
CAS nummer: 13368-42-8
UN: 2924
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %