Tetrabutylammonium dibromoiodide ≥97.0%

Leverantör: TCI
Varning

Synonyms: Tetrabutylammonium Dibromoiodide

T1269-1G T1269-10G
TCIAT1269-1GEA 470 SEK
TCIAT1269-1G TCIAT1269-10G
Tetrabutylammonium dibromoiodide ≥97.0%
Tetrabutylammonium dibromoiodide
Formel: C₁₆H₃₆Br₂IN
MW: 529.18 g/mol
MDL Number: MFCD00059121
CAS nummer: 15802-00-3

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (Iodometric Titration) min. 97.0 %
Melting point 79.0 to 83.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR