Vinyl trifluoroacetate ≥97.0% stabilised
Leverantör: TCI
Fara
Stabilization: med 4-tert-Butylpyrokatekol
T1216-25G T1216-5G
TCIAT1216-25GEA 21900 SEK
TCIAT1216-25G TCIAT1216-5G
Vinyl trifluoroacetate ≥97.0% stabilised
Vinyl trifluoroacetate
Formel: C₄H₃F₃O₂
Kokpunkt: 42°C
Flampunkt: –31°C
Storage Temperature: Kylskåp
MDL Number: MFCD00003561
CAS nummer: 433-28-3
EINECS: 207-088-1
UN: 2924
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %