Trimethylsilylmethyl azide ≥97.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
Azidomethyltrimethylsilane
T1184-1G
T1184-5G
TCIAT1184-1GEA
1150
SEK
TCIAT1184-1G
TCIAT1184-5G
Trimethylsilylmethyl azide ≥97.0%
Trimethylsilylmethyl azide
Formel:
C₄H₁₁N₃Si MW: 129,24 g/mol Kokpunkt: 60°C Flampunkt: 6°C |
MDL Number:
MFCD00059964 CAS nummer: 87576-94-1 UN: 1993 ADR: 3,II |
Specifikation testresultat
Appearance | Colorless to Light yellow to Red clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...