Tetrafluorotetracyanoquinodimethane ≥98.0% for organic electronics, purified by sublimation

Leverantör: TCI EUROPE
Fara

Synonyms: 2,3,5,6-Tetrafluoro-7,7,8,8-tetracyanoquinodimethane, TCNQF4, 2,2'-(Perfluorocyclohexa-2,5-diene-1,4-diylidene)dimalononitrile

T1131-1G T1131-100MG
TCIAT1131-1GEA 10525 SEK
TCIAT1131-1G TCIAT1131-100MG
Tetrafluorotetracyanoquinodimethane ≥98.0% for organic electronics, purified by sublimation
Tetrafluorotetracyanoquinodimethane
Formel: C₁₂F₄N₄
MW: 276,15 g/mol
Smältpunkt: 300°C
MDL Number: MFCD00042382
CAS nummer: 29261-33-4
UN: 3439
ADR: 6.1,III

Order Now

Specifikation testresultat

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR