Potassium monomethyl terephthalate ≥95.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: Terephthalic Acid Monomethyl Ester Potassium Salt

T1083-25G T1083-500G
TCIAT1083-25GEA 405 SEK
TCIAT1083-25G TCIAT1083-500G
Potassium monomethyl terephthalate ≥95.0% (by HPLC, titration analysis)
Potassium monomethyl terephthalate
Formel: C₉H₈O₄
MW: 218,25 g/mol
MDL Number: MFCD00058989
CAS nummer: 42967-55-5
EINECS: 256-027-5

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Ester Value) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR