1,2,3,4-Tetrahydrocarbazole 99,0 GC_ANALYSMETOD
Leverantör: TCI
|
Varning
|
Synonyms:
2,3,4,9-Tetrahydro-1H-carbazole
T1006-25G
T1006-100G
TCIAT1006-25GEA
1375
SEK
TCIAT1006-25G
TCIAT1006-100G
1,2,3,4-Tetrahydrocarbazole 99,0 GC_ANALYSMETOD
1,2,3,4-Tetrahydrocarbazole
Formel:
C₁₂H₁₃N MW: 171.24 g/mol Kokpunkt: 186°C |
MDL Number:
MFCD00004959 CAS nummer: 942-01-8 EINECS: 213-385-7 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 117.0 to 121.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...