Triethyl orthovalerate ≥95.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 1,1,1-Triethoxypentane, Orthovaleric Acid Triethyl Ester

T0977-25ML T0977-250ML
TCIAT0977-25MLEA 1625 SEK
TCIAT0977-25ML TCIAT0977-250ML
Triethyl orthovalerate ≥95.0%
Triethyl orthovalerate
Formel: C₁₁H₂₄O₃
MW: 204,31 g/mol
Kokpunkt: 50°C
MDL Number: MFCD00059383
CAS nummer: 919-29-9
EINECS: 213-047-9
UN: 3272
ADR: 3,III

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR