L-Theanine ≥98.0%

Leverantör: TCI EUROPE

Synonyms: (S)-2-Amino-5-(ethylamino)-5-oxopentanoic acid, L-gamma-Glutamylethylamide, N-gamma-Ethyl-L-glutamine

T0954-1G T0954-5G
TCIAT0954-1GEA 215 SEK
TCIAT0954-1G TCIAT0954-5G
L-Theanine ≥98.0%
L-Theanine
Formel: C₇H₁₄N₂O₃
MW: 174,20 g/mol
MDL Number: MFCD00059653
CAS nummer: 3081-61-6
EINECS: 221-379-0

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +7.6 to +8.4 deg (C=5, H2O)
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR