Trimethylolpropane triacrylate ≥75.0% stabilised

Leverantör: TCI
Varning

Synonyms: TMPTA, 1,1,1-Tris(acryloyloxymethyl)propane

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

T0949-25G T0949-500G
TCIAT0949-25GEA 245 SEK
TCIAT0949-25G TCIAT0949-500G
Trimethylolpropane triacrylate ≥75.0% stabilised
Trimethylolpropane triacrylate
Formel: C₁₅H₂₀O₆
MW: 296.32 g/mol
Flampunkt: 177°C
MDL Number: MFCD00008628
CAS nummer: 15625-89-5
EINECS: 239-701-3

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 75.0 %
Specific gravity (20/20) 1.1080 to 1.1180
Refractive index n20/D 1.4750 to 1.4770


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR