Triethylene glycol dimethacrylate ≥95.0% stabilised
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
(Ethane-1,2-diylbis(oxy))bis(ethane-2,1-diyl) bis(2-methylacrylate)
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
T0948-25ML
T0948-500ML
TCIAT0948-25MLEA
230
SEK
TCIAT0948-25ML
TCIAT0948-500ML
Triethylene glycol dimethacrylate ≥95.0% stabilised
Triethylene glycol dimethacrylate
Formel:
C₁₄H₂₂O₆ MW: 286,32 g/mol Kokpunkt: 155°C Flampunkt: 159°C |
MDL Number:
MFCD00008591 CAS nummer: 109-16-0 EINECS: 203-652-6 |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...