Triethylenetetramine-N,N,N',N'',N''',N'''-hexaacetic acid ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: TTHA

T0806-25G T0806-5G
TCIAT0806-25GEA 2825 SEK
TCIAT0806-25G TCIAT0806-5G
Triethylenetetramine-N,N,N',N'',N''',N'''-hexaacetic acid ≥98.0%
Triethylenetetramine-N,N,N',N'',N''',N'''-hexaacetic acid
Formel: C₁₈H₃₀N₄O₁₂
MW: 494,45 g/mol
MDL Number: MFCD00004290
CAS nummer: 869-52-3
EINECS: 212-790-6

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 %
Solubility in hot Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR