p-Tolyldiethanolamine ≥98.0%

Leverantör: TCI
Varning

T0797-25G T0797-500G T0797-100G
TCIAT0797-25GEA 295 SEK
TCIAT0797-25G TCIAT0797-500G TCIAT0797-100G
p-Tolyldiethanolamine ≥98.0%
p-Tolyldiethanolamine
Formel: C₁₁H₁₇NO₂
Kokpunkt: 163°C
MDL Number: MFCD00020580
CAS nummer: 3077-12-1
EINECS: 221-359-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc310ba02050709b4f8194b52272b297baa0ef65&vid=fc310ba02050709b4f8194b52272b297baa0ef65&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White or Colorles to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 98.0 %
Melting point 50.0 to 54.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR