1,2,3,10b-Tetrahydrofluoranthene 98,0 GC_ANALYSMETOD

Leverantör: TCI

T0771-1G T0771-5G
TCIAT0771-1GEA 1275 SEK
TCIAT0771-1G TCIAT0771-5G
1,2,3,10b-Tetrahydrofluoranthene 98,0 GC_ANALYSMETOD
1,2,3,10b-Tetrahydrofluoranthene
Formel: C₁₆H₁₄
MW: 206.29 g/mol
Kokpunkt: 205°C
MDL Number: MFCD00045348
CAS nummer: 20279-21-4
EINECS: 243-672-2

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 74.0 to 77.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR