2-Tridecenoic acid 80,0 GC_ANALYSMETOD
Leverantör: TCI
|
Varning
|
T0759-25G
T0759-5G
TCIAT0759-25GEA
2775
SEK
TCIAT0759-25G
TCIAT0759-5G
2-Tridecenoic acid 80,0 GC_ANALYSMETOD
2-Tridecenoic acid
Formel:
C₁₃H₂₄O₂ MW: 212.33 g/mol |
MDL Number:
MFCD00020487 CAS nummer: 6969-16-0 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to lump |
Purity (GC) | min. 80.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...